Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177772.1 | NP_001171243.1 | 4 | 754 | ||
NM_021471.2 | NP_067446.1 | 4 | 754 | |||
GenBank mRNA | AK045782.1 | - | 757 | |||
AK048926.1 | - | 4 | 724 | |||
AK161010.1 | - | 724 | ||||
AY007379.1 | - | 4 | 590 | |||
BC078456.1 | - | 4 | 704 | 30360663 |