Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021473.3 | NP_067448.1 | 5 | 737 | ||
GenBank mRNA | AF225564.1 | - | 5 | 642 | ||
AK005162.1 | - | 2 | 178 | |||
AK005376.1 | - | 5 | 672 | |||
AK009462.1 | - | 5 | 514 | |||
AK011157.1 | - | 2 | 178 | |||
AK011209.1 | - | 2 | 178 | |||
AK011221.1 | - | 2 | 177 | |||
AK011321.1 | - | 5 | 666 | |||
AK011388.1 | - | 5 | 692 | |||
AK011667.1 | - | 5 | 693 | |||
AK011794.1 | - | 5 | 666 | |||
AK011856.1 | - | 5 | 693 | |||
AK011906.1 | - | 5 | 671 | |||
AK011908.1 | - | 5 | 706 | |||
AK011918.1 | - | 5 | 692 | |||
AK128906.1 | - | 5 | 459 | |||
AK144198.1 | - | 5 | 693 | |||
AK146081.1 | - | 5 | 691 | |||
AK146278.1 | - | 5 | 693 | |||
AK152086.1 | - | 5 | 672 | |||
AK159573.1 | - | 5 | 691 | |||
AK160185.1 | - | 5 | 667 | |||
AK164461.1 | - | 5 | 692 | |||
AK168950.1 | - | 5 | 688 | |||
AK184360.1 | - | 1 | 38 | |||
AK187660.1 | - | 1 | 50 | |||
AK189553.1 | - | 1 | 38 | |||
AK196298.1 | - | 1 | 38 | |||
AK200998.1 | - | 1 | 25 | |||
AY207463.1 | - | 5 | 714 | |||
BC003850.1 | - | 5 | 521 | |||
BC039926.1 | - | 5 | 686 | 3156658 | ||
BC046762.1 | - | 5 | 643 | 5693736 |