Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080442.2 | NP_536690.1 | 1 | 227 | ||
GenBank mRNA | AF201734.1 | - | 1 | 192 | ||
AF298901.1 | - | 1 | 206 | |||
AK005983.1 | - | 1 | 219 | |||
AK019840.1 | - | 1 | 228 | |||
BC048470.1 | - | 1 | 224 | 6772177 |