Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163516.2 | NP_001156988.1 | 1, 2 | 351 | ||
NM_001289505.1 | NP_001276434.1 | 3, 4 | 329 | |||
NM_021483.4 | NP_067458.2 | 3, 4 | 461 | |||
GenBank mRNA | AB032591.1 | - | 3, 4 | 456 | ||
AK039764.1 | - | 3, 4 | 328 | |||
AK044552.1 | - | 3, 4 | 586 | |||
AK083141.1 | - | 1, 2 | 243 | |||
HQ646186.1 | - | 4, 5 | 680 | |||
HQ646187.1 | - | 3, 4 | 614 | |||
HQ646188.1 | - | 2, 3 | 509 | |||
HQ646189.1 | - | 2, 3 | 542 | |||
HQ646190.1 | - | 1, 2 | 437 | |||
HQ646191.1 | - | 3, 4 | 1270 | |||
HQ646192.1 | - | 4, 5 | 684 | |||
HQ646193.1 | - | 4, 5 | 590 | |||
HQ646194.1 | - | 3, 4 | 485 | |||
HQ646195.1 | - | 4, 5 | 458 | |||
HQ646196.1 | - | 3, 4 | 353 |