Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163516.2 | NP_001156988.1 | 4 | 696 | ||
NM_001289505.1 | NP_001276434.1 | 6 | 677 | |||
NM_021483.4 | NP_067458.2 | 6 | 806 | |||
GenBank mRNA | AB032591.1 | - | 6 | 801 | ||
AK039764.1 | - | 6 | 676 | |||
AK044552.1 | - | 6 | 934 | |||
AK083141.1 | - | 4 | 588 | |||
HQ646186.1 | - | 7 | 1025 | |||
HQ646187.1 | - | 6 | 959 | |||
HQ646188.1 | - | 5 | 854 | |||
HQ646189.1 | - | 5 | 887 | |||
HQ646190.1 | - | 4 | 782 | |||
HQ646191.1 | - | 6 | 1615 | |||
HQ646192.1 | - | 7 | 1029 | |||
HQ646193.1 | - | 7 | 935 | |||
HQ646194.1 | - | 6 | 830 | |||
HQ646195.1 | - | 7 | 803 | |||
HQ646196.1 | - | 6 | 698 |