Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153574.2 | NP_705802.1 | 5 | 537 | ||
GenBank mRNA | AJ510136.1 | - | 5 | 523 | ||
AK052421.1 | - | 5 | 478 | |||
AK054311.1 | - | 5 | 524 | |||
BC005554.1 | - | 5 | 442 | |||
BC054053.1 | - | 5 | 511 | |||
BC060978.1 | - | 5 | 474 | |||
BC080861.1 | - | 5 | 537 | 30357292 |