Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145937.3 | NP_666049.2 | 5 | 722 | ||
GenBank mRNA | AK151874.1 | - | 5 | 723 | ||
AK159192.1 | - | 5 | 711 | |||
AK160917.1 | - | 5 | 704 | |||
AK161203.1 | - | 5 | 715 | |||
AK216554.1 | - | 1 | 40 | |||
BC026981.1 | - | 5 | 695 | 5365580 |