Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033621.3 | NP_001028793.1 | 7 | 1136 | ||
GenBank mRNA | AF230979.2 | - | 7 | 1135 | ||
AK017447.1 | - | 7 | 1137 | |||
BC016214.1 | - | 7 | 1114 | 4482382 | ||
BC064052.1 | - | 7 | 1109 | 1366418 |