Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037085.1 | - | 3 | 196 | ||
BC049867.1 | - | 3 | 219 | |||
BC057921.1 | - | 3 | 207 | 5101105 | ||
CB948320.1 | - | 3 | 221 | |||
X99571.1 | - | 3 | 202 |