Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021500.2 | NP_067475.2 | 2 | 181 | ||
GenBank mRNA | AF263247.1 | - | 2 | 143 | ||
AK007307.1 | - | 2 | 152 | |||
AK034859.1 | - | 2 | 180 | |||
AK049424.1 | - | 2 | 201 | |||
AK075637.1 | - | 1 | 21 | |||
AK088854.1 | - | 2 | 189 | |||
AK145800.1 | - | 2 | 244 | |||
AK152523.1 | - | 2 | 189 | |||
AK168788.1 | - | 2 | 207 | |||
AK169149.1 | - | 2 | 189 | |||
BC039054.1 | - | 2 | 167 | 2648667 | ||
BC058687.1 | - | 2 | 179 | 6411707 | ||
CX212584.1 | - | 2 | 181 |