Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042491.2 | NP_001035956.1 | 10 | 1362 | ||
NM_001289517.1 | NP_001276446.1 | 10 | 1362 | |||
NM_001289518.1 | NP_001276447.1 | 10 | 1401 | |||
NM_001289519.1 | NP_001276448.1 | 10 | 1401 | |||
NM_001289520.1 | NP_001276449.1 | 10 | 1377 | |||
NM_021505.3 | NP_067480.2 | 10 | 1401 | |||
GenBank mRNA | AB041593.1 | - | 10 | 1345 | ||
AK012579.1 | - | 3 | 301 | |||
AK031432.1 | - | 10 | 1326 | |||
AK049719.1 | - | 10 | 1355 | |||
AK075969.1 | - | 10 | 1338 | |||
AK088327.1 | - | 10 | 1364 | |||
AK147951.1 | - | 9 | 2838 | |||
AK148679.1 | - | 10 | 1317 | |||
AK151314.1 | - | 10 | 1308 | |||
AK151905.1 | - | 10 | 1347 | |||
AK152528.1 | - | 10 | 1278 | |||
AK152981.1 | - | 10 | 1309 | |||
AK153373.1 | - | 10 | 1308 | |||
AK153422.1 | - | 10 | 1308 | |||
AK159204.1 | - | 10 | 1308 | |||
AK159713.1 | - | 10 | 1316 | |||
AK159916.1 | - | 10 | 1340 | |||
AK167561.1 | - | 10 | 1301 | |||
AK168135.1 | - | 10 | 1325 | |||
AK168737.1 | - | 10 | 1302 | |||
AK168814.1 | - | 10 | 1340 | |||
AK169196.1 | - | 10 | 1364 | |||
AK178093.1 | - | 3 | 244 | |||
AK180885.1 | - | 3 | 244 | |||
AK182176.1 | - | 3 | 244 | |||
AK183979.1 | - | 3 | 206 | |||
AK184544.1 | - | 3 | 205 | |||
AK185729.1 | - | 3 | 206 | |||
BC010339.1 | - | 10 | 1317 | 2648173 | ||
BC046566.1 | - | 10 | 1341 | 2650790 | ||
BC046804.1 | - | 10 | 1299 | 6404923 |