Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042491.2 | NP_001035956.1 | 12 | 1589 | ||
NM_001289517.1 | NP_001276446.1 | 12 | 1589 | |||
NM_001289518.1 | NP_001276447.1 | 12 | 1628 | |||
NM_001289519.1 | NP_001276448.1 | 12 | 1628 | |||
NM_001289520.1 | NP_001276449.1 | 12 | 1604 | |||
NM_021505.3 | NP_067480.2 | 12 | 1628 | |||
GenBank mRNA | AB041593.1 | - | 12 | 1572 | ||
AK012579.1 | - | 5 | 528 | |||
AK031432.1 | - | 12 | 1553 | |||
AK049719.1 | - | 12 | 1582 | |||
AK075969.1 | - | 12 | 1565 | |||
AK088327.1 | - | 12 | 1591 | |||
AK148679.1 | - | 12 | 1544 | |||
AK151314.1 | - | 12 | 1535 | |||
AK151905.1 | - | 12 | 1574 | |||
AK152528.1 | - | 12 | 1505 | |||
AK152981.1 | - | 12 | 1536 | |||
AK153373.1 | - | 12 | 1535 | |||
AK153422.1 | - | 12 | 1535 | |||
AK159204.1 | - | 12 | 1535 | |||
AK159713.1 | - | 12 | 1543 | |||
AK159916.1 | - | 12 | 1567 | |||
AK167561.1 | - | 11 | 2504 | |||
AK168135.1 | - | 12 | 1552 | |||
AK168737.1 | - | 12 | 1529 | |||
AK168814.1 | - | 12 | 1567 | |||
AK169196.1 | - | 12 | 1591 | |||
AK209720.1 | - | 1 | 31 | |||
BC010339.1 | - | 12 | 1544 | 2648173 | ||
BC046566.1 | - | 12 | 1568 | 2650790 | ||
BC046804.1 | - | 12 | 1526 | 6404923 |