Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042491.2 | NP_001035956.1 | 16 | 2027 | ||
NM_001289519.1 | NP_001276448.1 | 16 | 2046 | |||
NM_001289520.1 | NP_001276449.1 | 16 | 2042 | |||
NM_021505.3 | NP_067480.2 | 16 | 2066 | |||
GenBank mRNA | AB041593.1 | - | 16 | 2010 | ||
AK012579.1 | - | 9 | 966 | |||
AK031432.1 | - | 16 | 1991 | |||
AK049719.1 | - | 16 | 2020 | |||
AK075969.1 | - | 16 | 1983 | |||
AK088327.1 | - | 16 | 2029 | |||
AK148679.1 | - | 16 | 1982 | |||
AK152528.1 | - | 16 | 1942 | |||
AK159204.1 | - | 16 | 1973 | |||
AK159713.1 | - | 16 | 1981 | |||
AK159916.1 | - | 16 | 2005 | |||
AK167561.1 | - | 15 | 2942 | |||
AK168135.1 | - | 16 | 1990 | |||
AK168737.1 | - | 16 | 1967 | |||
AK168814.1 | - | 16 | 2005 | |||
BC010339.1 | - | 16 | 1982 | 2648173 | ||
BC011386.1 | - | 3 | 199 | |||
BC046804.1 | - | 16 | 1964 | 6404923 |