Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021506.2 | NP_067481.2 | 7, 8 | 1639 | ||
GenBank mRNA | AF030131.1 | - | 7, 8 | 1541 | ||
AK045470.1 | - | 5, 6 | 774 | |||
AK148137.1 | - | 7, 8 | 1570 | |||
AK173185.1 | - | 7, 8 | 2005 | |||
BC060113.1 | - | 8, 9 | 1511 | 6405370 | ||
BC060696.1 | - | 8, 9 | 1511 | 6405370 |