Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021510.2 | NP_067485.1 | 4 | 423 | ||
GenBank mRNA | AK052983.1 | - | 4 | 377 | ||
AK088169.1 | - | 4 | 376 | |||
AK161491.1 | - | 521 | ||||
AK178902.1 | - | 1 | 30 | |||
BC016459.1 | - | 4 | 391 | |||
BC027002.1 | - | 3 | 336 | |||
BC042187.1 | - | 4 | 358 | 5376052 | ||
BC056224.1 | - | 3 | 353 | 6515270 | ||
BU058854.1 | - | 2 | 176 | |||
Y14196.1 | - | 4 | 381 |