Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021512.2 | NP_067487.1 | 28 | 3772 | ||
GenBank mRNA | AF104415.2 | - | 28 | 3772 | ||
AK172911.1 | - | 14 | 1498 | |||
BC052450.1 | - | 27 | 3130 | 6409240 | ||
BC054523.1 | - | 27 | 3130 | 6409240 |