Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146001.1 | NP_001139473.1 | 5 | 781 | ||
NM_021517.2 | NP_067492.2 | 5 | 827 | |||
GenBank mRNA | AF220100.1 | - | 5 | 766 | ||
AF474247.1 | - | 4 | 766 | |||
AK006269.1 | - | 5 | 1301 | |||
AK012070.1 | - | 5 | 817 | |||
AK014879.1 | - | 5 | 1084 | |||
AK029752.1 | - | 5 | 1285 | |||
AK029764.1 | - | 5 | 1295 | |||
AK165415.1 | - | 5 | 762 | |||
BC013512.1 | - | 4 | 750 | 4234949 |