Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021518.3 | NP_067493.1 | 6 | 604 | ||
GenBank mRNA | AB232585.1 | - | 6 | 385 | ||
AK042862.1 | - | 6 | 574 | |||
AK079071.1 | - | 6 | 605 | |||
AK135421.1 | - | 6 | 588 | |||
AK146179.1 | - | 6 | 589 | |||
AK150030.1 | - | 6 | 636 | |||
AK159607.1 | - | 6 | 625 | |||
AK160333.1 | - | 6 | 605 | |||
AK163320.1 | - | 6 | 587 | |||
AK181047.1 | - | 2 | 108 | |||
AK184956.1 | - | 2 | 108 | |||
AK208722.1 | - | 2 | 108 | |||
AK212465.1 | - | 2 | 108 | |||
AK212917.1 | - | 2 | 130 | |||
AK216186.1 | - | 2 | 108 | |||
BC058382.1 | - | 6 | 602 | 6820798 | ||
X95403.1 | - | 6 | 592 |