Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021526.2 | NP_067501.2 | 7, 8 | 767 | ||
GenBank mRNA | AK011974.1 | - | 7, 8 | 716 | ||
AK012013.1 | - | 7, 8 | 698 | |||
AK031863.1 | - | 7, 8 | 704 | |||
AK078134.1 | - | 7, 8 | 719 | |||
AK150280.1 | - | 7, 8 | 703 | |||
AK151104.1 | - | 7, 8 | 750 | |||
AK151733.1 | - | 7, 8 | 750 | |||
BC003742.1 | - | 7, 8 | 672 | 3599671 | ||
Y13071.1 | - | 7, 8 | 688 |