Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021529.3 | NP_067504.2 | 7 | 759 | ||
GenBank mRNA | AB041577.1 | - | 7 | 748 | ||
AJ238247.2 | - | 6 | 586 | |||
AK013403.1 | - | 6 | 555 | |||
AK015746.1 | - | 6 | 588 | |||
AK028063.1 | - | 7 | 724 | |||
AK028254.1 | - | 6 | 678 | |||
AK033405.1 | - | 7 | 758 | |||
AK040589.1 | - | 6 | 587 | |||
AK087572.1 | - | 6 | 582 | |||
AK088240.1 | - | 6 | 668 | |||
AK163192.1 | - | 758 | ||||
AK168298.1 | - | 7 | 761 | |||
BC024754.1 | - | 7 | 722 | 5375405 | ||
BC138295.1 | - | 6 | 555 | 8861314 | ||
BC138296.1 | - | 6 | 555 | 8861315 |