Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021531.6 | NP_067506.2 | 14 | 1721 | ||
NM_153141.1 | NP_694781.1 | 14 | 1721 | |||
GenBank mRNA | AF117887.2 | - | 14 | 1631 | ||
AK158242.1 | - | 14 | 1686 | |||
AK158757.1 | - | 14 | 1561 | |||
AK189449.1 | - | 1 | 30 | |||
BC003964.1 | - | 9 | 828 | |||
BC008263.1 | - | 4 | 327 | |||
BC026427.1 | - | 4 | 287 | |||
BC036974.1 | - | 14 | 1705 | 4935077 | ||
BC071267.1 | - | 15 | 1837 |