Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190466.1 | NP_001177395.1 | 4 | 1246 | ||
NM_021532.4 | NP_067507.2 | 4 | 1135 | |||
GenBank mRNA | AF488775.1 | - | 4 | 1035 | ||
AK077691.1 | - | 4 | 752 | |||
AY208970.1 | - | 4 | 987 | |||
BC052999.1 | - | 2 | 444 | |||
BC145833.1 | - | 4 | 1085 | 40131136 |