Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190466.1 | NP_001177395.1 | 4 | 875 | ||
NM_021532.4 | NP_067507.2 | 4 | 764 | |||
GenBank mRNA | AF251078.1 | - | 2 | 100 | ||
AF488775.1 | - | 4 | 664 | |||
AK077691.1 | - | 4 | 381 | |||
AK199966.1 | - | 2 | 90 | |||
AY208970.1 | - | 4 | 616 | |||
BC052999.1 | - | 2 | 73 | |||
BC145833.1 | - | 4 | 714 | 40131136 |