Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163354.1 | NP_001156826.1 | 14 | 1346 | ||
NM_001163355.1 | NP_001156827.1 | 14 | 1346 | |||
NM_021536.7 | NP_067511.4 | 14 | 1346 | |||
GenBank mRNA | AK019059.1 | - | 14 | 1306 | ||
AK028597.1 | - | 14 | 1289 | |||
AK031139.1 | - | 12 | 1008 | |||
AK032096.1 | - | 14 | 1319 | |||
AK041119.1 | - | 14 | 1282 | |||
AK043323.1 | - | 5 | 403 | |||
AK054472.1 | - | 14 | 1317 | |||
AK078955.1 | - | 14 | 1289 | |||
AK090071.1 | - | 7 | 672 | |||
AK138830.1 | - | 14 | 1287 | |||
AK141621.1 | - | 14 | 1173 | |||
AK147749.1 | - | 14 | 1325 | |||
AK147801.1 | - | 14 | 1351 | |||
AK167502.1 | - | 14 | 1346 | |||
AK169811.1 | - | 13 | 2003 | |||
BC006918.1 | - | 5 | 437 | |||
BC044301.1 | - | 3 | 185 | |||
BC046785.1 | - | 14 | 1329 | 6416127 | ||
BC052882.1 | - | 4 | 290 | |||
BC058350.1 | - | 14 | 1261 | 6815832 |