Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021547.3 | NP_067522.1 | 15 | 1440 | ||
GenBank mRNA | AK041538.1 | - | 15 | 1396 | ||
AK150166.1 | - | 15 | 1487 | |||
AK150254.1 | - | 15 | 1487 | |||
AK155124.1 | - | 15 | 1379 | |||
AK160480.1 | - | 14 | 2334 | |||
BC003313.1 | - | 15 | 1357 | 3155179 | ||
BC058727.1 | - | 16 | 1527 | |||
X82457.1 | - | 15 | 1424 |