Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021551.4 | NP_067526.2 | 5, 6 | 1232 | ||
GenBank mRNA | AK007059.1 | - | 5, 6 | 1177 | ||
BC062878.1 | - | 6, 7 | 1094 | 5696414 | ||
DQ846835.1 | - | 2, 3 | 169 |