Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021554.2 | NP_067529.2 | 2, 3 | 715 | ||
GenBank mRNA | AB041664.1 | - | 2, 3 | 493 | ||
AJ278576.1 | - | 2, 3 | 715 | |||
AK075586.1 | - | 2, 3 | 478 | |||
AK169951.1 | - | 1, 2 | 131 | |||
AK201397.1 | - | 2, 3 | 204 | |||
BC023188.1 | - | 1, 2 | 156 | |||
BC068124.1 | - | 2, 3 | 509 | 5716972 | ||
BC082317.1 | - | 1, 2 | 140 |