Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021554.2 | NP_067529.2 | 3 | 843 | ||
GenBank mRNA | AB041664.1 | - | 3 | 620 | ||
AJ278576.1 | - | 3 | 843 | |||
AK075586.1 | - | 3 | 606 | |||
AK169951.1 | - | 2 | 259 | |||
AK188768.1 | - | 1 | 53 | |||
AK202674.1 | - | 1 | 53 | |||
AK206681.1 | - | 1 | 45 | |||
AK209406.1 | - | 1 | 53 | |||
AK210173.1 | - | 1 | 53 | |||
AK217664.1 | - | 1 | 53 | |||
AK219043.1 | - | 1 | 53 | |||
BC023188.1 | - | 2 | 284 | |||
BC068124.1 | - | 3 | 637 | 5716972 | ||
BC082317.1 | - | 2 | 268 |