Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204426.1 | NP_001191355.1 | 3 | 333 | ||
NM_002314.3 | NP_002305.1 | 4 | 555 | |||
GenBank mRNA | AB209557.1 | - | 1 | 27 | ||
AF134379.1 | - | 4 | 341 | |||
AK290162.1 | - | 5 | 480 | |||
AK300382.1 | - | 3 | 272 | |||
D26309.1 | - | 5 | 436 | |||
DC296332.1 | - | 3 | 333 |