Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204426.1 | NP_001191355.1 | 5 | 623 | ||
NM_002314.3 | NP_002305.1 | 6 | 845 | |||
GenBank mRNA | AB209557.1 | - | 3 | 317 | ||
AF134379.1 | - | 6 | 631 | |||
AK290162.1 | - | 7 | 770 | |||
AK300382.1 | - | 5 | 562 | |||
D26309.1 | - | 7 | 726 |