Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021567.5 | NP_067542.3 | 7 | 736 | ||
GenBank mRNA | AF176328.1 | - | 7 | 622 | ||
AK004580.1 | - | 7 | 687 | |||
AK005383.1 | - | 7 | 685 | |||
AK077632.1 | - | 7 | 686 | |||
AK082005.1 | - | 7 | 686 | |||
BC010694.1 | - | 7 | 625 | 2654291 |