Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021605.3 | NP_067618.1 | 10 | 1028 | ||
GenBank mRNA | AF217650.1 | - | 9 | 879 | ||
AK035502.1 | - | 10 | 1027 | |||
AK088173.1 | - | 10 | 1077 | |||
AK149308.1 | - | 10 | 1076 | |||
AK165583.1 | - | 8 | 797 | |||
BC026541.1 | - | 10 | 1018 | |||
BC037697.1 | - | 10 | 1005 | 3994871 |