Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ackr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276719.1 | NP_001263648.1 | 4 | 868 | ||
NM_021609.4 | NP_067622.2 | 3 | 756 | |||
GenBank mRNA | AK028281.1 | - | 3 | 648 | ||
AK145960.1 | - | 4 | 753 | |||
AK147166.1 | - | 4 | 760 | |||
AK157140.1 | - | 4 | 869 | |||
BC113146.1 | - | 2 | 489 | 40055448 | ||
Y12879.1 | - | 2 | 559 |