Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC125528.1 | - | 3 | 321 | ||
CJ236701.1 | - | 3 | 138 | |||
X65979.1 | - | 2 | 146 | |||
X65981.1 | - | 3 | 354 |