Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK185752.1 | - | 285 |
Gene Symbol: Wbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021714.4 | NP_068360.4 | 6 | 696 | ||
GenBank mRNA | AK031678.1 | - | 6 | 521 | ||
AK041079.1 | - | 6 | 635 | |||
AK144968.1 | - | 6 | 620 | |||
AK145049.1 | - | 6 | 624 | |||
AK145917.1 | - | 6 | 646 | |||
AK151989.1 | - | 6 | 642 | |||
AK153044.1 | - | 6 | 642 | |||
AK159257.1 | - | 6 | 634 | |||
AK205178.1 | - | 1 | 21 | |||
BC006600.1 | - | 6 | 608 | 3156999 | ||
BC021823.1 | - | 6 | 589 | 3994221 |