Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fign | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267846.1 | NP_001254775.1 | 4 | 2147 | ||
NM_001267847.1 | NP_001254776.1 | 4 | 2128 | |||
NM_021716.5 | NP_068362.1 | 3 | 2089 | |||
GenBank mRNA | AF263913.1 | - | 3 | 2039 | ||
AK143787.1 | - | 3 | 2064 | |||
AK163474.1 | - | 3 | 2060 | |||
AK164536.1 | - | 4 | 2100 | |||
BC059266.1 | - | 4 | 2147 | 6409940 |