Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024177.3 | NP_077139.2 | 2, 3 | 345 | ||
GenBank mRNA | AK088343.1 | - | 2, 3 | 284 | ||
AK151598.1 | - | 2, 3 | 284 | |||
BC002319.1 | - | 1, 2 | 321 | 3593984 | ||
BC030739.1 | - | 2, 3 | 272 | 1493919 |