Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291041.1 | NP_001277970.1 | 4 | 442 | ||
NM_021782.3 | NP_068554.1 | 4 | 442 | |||
GenBank mRNA | AF254070.1 | - | 4 | 424 | ||
DQ645418.1 | - | 4 | 371 |