Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019811.3 | NP_062785.2 | 7 | 812 | ||
GenBank mRNA | AF216873.1 | - | 7 | 762 | ||
AK028586.1 | - | 7 | 790 | |||
AK029014.1 | - | 7 | 780 | |||
AK029262.1 | - | 798 | ||||
AK053877.1 | - | 7 | 786 | |||
AK086921.1 | - | 3 | 1607 | |||
AK093831.1 | - | 7 | 789 | |||
BC040790.1 | - | 7 | 762 | |||
BC051432.1 | - | 7 | 812 | 6515568 |