Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021890.3 | NP_068690.3 | 9 | 1205 | ||
GenBank mRNA | AB041560.1 | - | 9 | 1147 | ||
AK029318.1 | - | 9 | 1064 | |||
AK080414.1 | - | 9 | 1057 | |||
AK154534.1 | - | 9 | 1082 | |||
AK172509.1 | - | 9 | 1184 | |||
BC055950.1 | - | 9 | 886 | |||
BC071266.1 | - | 8 | 1193 |