Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fignl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163359.1 | NP_001156831.1 | 3 | 909 | ||
NM_001163360.1 | NP_001156832.1 | 2 | 779 | |||
NM_021891.3 | NP_068691.2 | 3 | 899 | |||
GenBank mRNA | AF263914.1 | - | 2 | 813 | ||
AK051324.1 | - | 3 | 1007 | |||
AK051874.1 | - | 3 | 869 | |||
AK088574.1 | - | 3 | 882 | |||
AK143850.1 | - | 3 | 878 | |||
AK148994.1 | - | 1 | 932 | |||
BC051942.1 | - | 3 | 852 | 6401467 | ||
BC052415.1 | - | 3 | 852 | 6401467 |