Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113532.1 | NP_001107004.1 | 6 | 604 | ||
NM_001113533.1 | NP_001107005.1 | 6 | 604 | |||
NM_175394.2 | NP_780603.1 | 6 | 652 | |||
GenBank mRNA | AJ276707.1 | - | 4 | 362 | ||
AK031965.1 | - | 6 | 661 | |||
AK076111.1 | - | 6 | 646 | |||
AK156334.1 | - | 6 | 500 | |||
AK220249.1 | - | 2 | 259 | |||
BC046416.1 | - | 6 | 533 | |||
BC047338.1 | - | 6 | 533 | |||
BC093504.1 | - | 6 | 537 |