Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021921.3 | NP_068740.3 | 3, 4 | 767 | ||
GenBank mRNA | AF220195.1 | - | 3, 4 | 486 | ||
AF310135.1 | - | 3, 4 | 548 | |||
AK014339.1 | - | 3, 4 | 471 | |||
AK158968.1 | - | 3, 4 | 621 | |||
BC029704.1 | - | 3, 4 | 546 | 5359447 |