Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005357.3 | NP_005348.2 | 1 | 805 | ||
XM_005258937.3 | XP_005258994.1 | 1 | 800 | |||
GenBank mRNA | AK292482.1 | - | 1 | 785 | ||
AK309893.1 | - | 1 | 728 | |||
BC029961.1 | - | 1 | 792 | |||
BC070041.1 | - | 1 | 805 | 5296155 |