Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022022.3 | NP_071305.2 | 12 | 2134 | ||
GenBank mRNA | AF260926.1 | - | 12 | 2249 | ||
AK029914.1 | - | 12 | 1739 | |||
AK122345.1 | - | 12 | 1645 | |||
BC067402.1 | - | 12 | 1881 | 30099547 | ||
BC075620.1 | - | 12 | 2095 | 6405514 |