Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 11 | 1142 | ||
NM_001199114.1 | NP_001186043.1 | 10 | 981 | |||
NM_001199115.1 | NP_001186044.1 | 10 | 1081 | |||
NM_001199116.1 | NP_001186045.1 | 10 | 1041 | |||
NM_022880.3 | NP_075018.1 | 9 | 1082 | |||
GenBank mRNA | AA407561.1 | - | 4 | 248 | ||
AF131212.1 | - | 9 | 1083 | |||
AF305501.1 | - | 10 | 1052 | |||
AK004756.1 | - | 10 | 1027 | |||
AK050089.1 | - | 10 | 1066 | |||
AK146265.1 | - | 10 | 982 | |||
AK149483.1 | - | 10 | 997 | |||
AK170463.1 | - | 10 | 1013 | |||
AK198320.1 | - | 1 | 28 | |||
AK212529.1 | - | 1 | 28 | |||
BC004828.1 | - | 11 | 1105 | 2631728 | ||
BC006812.1 | - | 9 | 1013 | 3155188 | ||
EU180577.1 | - | 9 | 957 |