Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 4, 5 | 352 | ||
NM_001199114.1 | NP_001186043.1 | 3, 4 | 191 | |||
NM_001199115.1 | NP_001186044.1 | 3, 4 | 297 | |||
NM_001199116.1 | NP_001186045.1 | 3, 4 | 257 | |||
NM_022880.3 | NP_075018.1 | 2, 3 | 298 | |||
GenBank mRNA | AF131212.1 | - | 2, 3 | 299 | ||
AF305501.1 | - | 3, 4 | 262 | |||
AK004756.1 | - | 3, 4 | 243 | |||
AK050089.1 | - | 3, 4 | 282 | |||
AK146265.1 | - | 3, 4 | 192 | |||
AK149483.1 | - | 3, 4 | 213 | |||
AK170463.1 | - | 3, 4 | 229 | |||
AK177360.1 | - | 291 | ||||
AK178838.1 | - | 125 | ||||
AK180131.1 | - | 289 | ||||
AK197634.1 | - | 289 | ||||
AK205425.1 | - | 288 | ||||
AK218485.1 | - | 179 | ||||
BC004828.1 | - | 4, 5 | 315 | 2631728 | ||
BC006812.1 | - | 2, 3 | 229 | 3155188 | ||
BY055687.1 | - | 3, 4 | 192 | |||
CD540408.1 | - | 2, 3 | 217 | |||
CX219686.1 | - | 4, 5 | 347 |