Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199113.1 | NP_001186042.1 | 5 | 398 | ||
NM_001199114.1 | NP_001186043.1 | 4 | 237 | |||
NM_001199115.1 | NP_001186044.1 | 4 | 343 | |||
NM_001199116.1 | NP_001186045.1 | 4 | 303 | |||
NM_022880.3 | NP_075018.1 | 3 | 344 | |||
GenBank mRNA | AF131212.1 | - | 3 | 345 | ||
AF305501.1 | - | 4 | 308 | |||
AK050089.1 | - | 4 | 328 | |||
AK146265.1 | - | 4 | 238 | |||
AK149483.1 | - | 4 | 259 | |||
AK170463.1 | - | 4 | 275 | |||
BB853265.1 | - | 5 | 408 | |||
BC004828.1 | - | 5 | 361 | 2631728 | ||
BC006812.1 | - | 3 | 275 | 3155188 | ||
BY055687.1 | - | 4 | 238 | |||
CD540408.1 | - | 3 | 263 | |||
CX219686.1 | - | 5 | 393 | |||
EU180577.1 | - | 3 | 213 |