Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022023.2 | NP_071306.2 | 2, 3 | 245 | ||
GenBank mRNA | AB001732.1 | - | 2, 3 | 177 | ||
AB050013.1 | - | 2, 3 | 187 | |||
AF297220.1 | - | 2, 3 | 223 | |||
AK013947.1 | - | 2, 3 | 202 | |||
AK013970.1 | - | 2, 3 | 198 | |||
AK142014.1 | - | 2, 3 | 217 | |||
BC040233.1 | - | 2, 3 | 179 | 5364203 | ||
BC049134.1 | - | 2, 3 | 183 | 6831501 |