Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026570.4 | NP_080846.1 | 3, 4 | 436 | ||
GenBank mRNA | AK002839.1 | - | 2, 3 | 84 | ||
AK010522.1 | - | 3, 4 | 437 | |||
AK012096.1 | - | 3, 4 | 403 | |||
AK182917.1 | - | 2, 3 | 89 | |||
AK219066.1 | - | 2, 3 | 89 | |||
BC020043.1 | - | 3, 4 | 404 | 4013548 |